Sodium sulphite, anhydrous 98,0 Iodometry, powder, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

3922-01EA 86 GBP
3922-01 3922-05 3922-07
Sodium sulphite, anhydrous 98,0 Iodometry, powder, BAKER ANALYZED® ACS, J.T.Baker®
Sodium sulphite
Formula: Na₂SO₃
MW: 126.04 g/mol
Melting Pt: > 500 °C
Density: 2.63 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003503
CAS Number: 7757-83-7
EINECS: 231-821-4
REACH: 01-2119537420-49
Merck Index: 13,08757

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use = 98.0 %
Assay (Na₂SO₃) (by iodometry) ≥98.0 %
Insoluble Matter ≤0.005 %
Free Acid Passes Test
Titrable Free Base (meq/g) ≤0.03
Chloride (Cl) ≤0.02 %
Heavy Metals (as Pb) ≤0.001 %
Trace Impurities - Iron (Fe) ≤0.001 %
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR