Cell culture plates, Advanced TC™
Supplier: Greiner Bio-One
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
657960_100 665980_100 677980_100 662960_100
392-0231EA 533 GBP
392-0231 392-0246 392-0249 392-0242
Cell culture plates, Advanced TC™
Microplates Cell Culture Plates
Microplates with a modified culture surface to optimise cell characteristics and functions. The improved adherence and increased growth rate improve and accelerate cell proliferation. Additionally, the Advanced TC™ surface facilitates uniform and consistent cell adherence, increases throughput and reduces cell loss, for example during automated washing stages. Free of detectable DNase, RNase and human DNA.
  • Increased cell adherence
  • Morphology similar to in vivo conditions
  • Optimal cultivation conditions for sensitive cells
  • Enables reduced quantities of serum
More Product Information