Microplates, 1536-well
Supplier: Greiner Bio-One
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
782061_60 782074_60 782077_60 783096_60 783076_60 782075_60 782076_60 782095_60 782096_60 782101_60
392-0138EA 2300 GBP
392-0138 738-0067 738-0070 738-0089 738-0084 738-0068 738-0069 738-0077 738-0078 738-0080
Microplates, 1536-well
Microplates HTS Microplates
PS, without lid. Ideal for high throughput screening applications, these plates are available as clear bottom variants (in clear PS) and black or white in LUMITRAC™ or FLUOTRAC™ quality.
  • HiBase versions designed for top reading instruments, µClear® LoBase plates for bottom reading instruments
  • Solid bottom plates available with high binding (MICROLON™ 600, LUMITRAC™ 600 or FLUOTRAC™ 600) or medium binding (LUMITRAC™ 200 or FLUOTRAC™ 200) surfaces
  • Rounded square well design combines the advantages of the square well (flexible working volume, 3 to 10 µl)
More Product Information