Iodine monochloride

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Iodochloride

39104.22 39104.36 39104.14
39104.22EA 57.4 GBP
39104.22 39104.36 39104.14
Iodine monochloride
Iodine monochloride

CAS No.: 7790-99-0 Molecular Formula: ClI Formula Weight: 162.36 Physical Form: Crystalline/Liquid Density: 3.1822 Sensitivity: Moisture and Light Sensitive Appearance: Dark red/black Odour: Pungent Melting Point: 25-27° Boiling Point: 96-98° Solubility: Soluble in water, alcohol, ether, carbon disulfide Beilstein Reference: 3902972 Merck Reference: 14,5017 UN No.: 1792 MDL No.: MFCD00011354

Formula: ClI
MW: 162.36 g/mol
Boiling Pt: 96…98 °C
Melting Pt: 25…27 °C
Density: 3.1822
Storage Temperature: Ambient
MDL Number: MFCD00011354
CAS Number: 7790-99-0
EINECS: 232-236-7
UN: 1792
ADR: 8 II
Merck Index: 13,05039

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR