VWR®, Pierceable Films for ELISA and General Incubation

Supplier: VWR Collection
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

391-1263EA 76.2 GBP
391-1263 391-1277
VWR®, Pierceable Films for ELISA and General Incubation
Seals Microplate Seals
Easily pierceable 94,1 µm thick PE films with acrylic adhesive allow direct sample recovery with singe- or multi-channel pipettes and robotic probes.

  • Functional temperature range from −40 to 90 °C

Each film L×D: 142,9×82,6 mm

Each film L×D: 142,9×82,6 mm
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR