VWR® Petri Dishes, Glass
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
391-0652EA 34.5 GBP
391-0652 391-0659 391-0660 391-0665 391-0666 391-0667 391-0668 391-0669 391-0671 391-0673 391-0674 391-0698
VWR® Petri Dishes, Glass
Culture Dishes Petri Dishes
Soda-lime glass, with lid. For use as disposables. These Petri dishes are a green alternative to disposable Petri dishes made from plastic.
  • Good chemical resistance
  • Heatable up to 60 °C
  • Not suitable for applications related to strong heat or rapid temperature changes
  • Ideal for bacterial culture production
Caution: The indication of the diameter of the petri dishes is approximate.