Caps, KIM-KAP™, disposable, KIMBLE®
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SP73660-13 SP73660-16 SP73660-18 SP73660-20 SP73660-25 SP73660-38 SP73662-13 SP73662-16 SP73662-18 SP73662-20 SP73662-25 SP73663-13 SP73663-16 SP73663-18 SP73663-20 SP73663-25 SP73664-13 SP73664-16 SP73664-18 SP73664-20 SP73664-25 SP73665-13 SP73665-16 SP73665-18 SP73665-20
391-0495EA 31.3 GBP
391-0495 391-0496 391-0497 391-0498 391-0499 391-0500 391-0501 391-0503 391-0505 391-0507 391-0508 391-0509 391-0510 391-0511 391-0512 391-0513 391-0514 391-0515 391-0516 391-0517 391-0518 391-0519 391-0520 391-0521 391-0523
Caps, KIM-KAP™, disposable, KIMBLE®
Closures Test/Sample Tube Closures
PP, coloured.
These caps are designed to fit culture tubes with outer Ø ranging from 13 to 38 mm.