Petri dishes
Supplier: Greiner Bio-One
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
633180_480 633181_480 639102_120 632181 639161_120 632180_480 627102_740 664102_360 628102_600 635102_480 628161_600 627161_740 664161_360 635161_480
391-3660EA 128 GBP
391-3660 391-0490 391-3665 391-0488 391-0491 391-3661 391-2072 391-2079 391-3655 391-0261 391-0487 391-0486 391-0493 391-0558
Petri dishes
Culture Dishes Petri Dishes
Made from crystal clear PS for best possible use in microscopic applications. Versions with or without vents.
  • Easily stackable
  • Heavy construction for good thermal resistance when working with hot agar
  • With vents for better gas exchange or without vents for long incubation times