Inoculating loops, Microstreaker
Supplier: MEDICAL WIRE EQUIPMENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MW180 MW182 MW184
300-0501EA 97.2 GBP
300-0501 391-0471 391-0472
Inoculating loops, Microstreaker
Spreaders Inoculating Loops and Needles
Inoculation loop of nickel-chromium wire of not more than 60 mm length, conforming to Howie recommendations. Colour coded handle.