Constant climate chambers with Peltier technology, KBF-S ECO Solid.Line series
Supplier: Binder
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9020-0416 9020-0418 9020-0419 9020-0463
390-1594EA 11630 GBP
390-1594 390-1595 390-1596 390-1696
Constant climate chambers with Peltier technology, KBF-S ECO Solid.Line series
Testing Chambers Climatic Chambers
The KBF-S ECO climatic chambers are very energy-efficient thanks to thermoelectric cooling technology with heat dissipation. The thermoelectric cooling technology means the KBF-S ECO series is also very quiet during operation.
  • APT.line™ preheating chamber technology
  • Independent water supply via tank
  • LCD shows temperature and humidity along with additional information and alarms
  • Internal data logger, measured values can be read out in open format via USB
  • Unit self-test for comprehensive status analysis
  • Inner chamber made completely of stainless steel
  • Access
More Product Information