CO₂ incubators, MCO series, PHCbi
Supplier: PHC Corporation
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MCO-230AIC-PE MCO-170-AC-PE
390-0979EA 7970 GBP
390-0979 SANYMCO-170-AC-PE
CO₂ incubators, MCO series, PHCbi
Incubators CO₂ Incubators
MCO incubators maintain a constant germicidal protection with inCu saFe™ a copper-enriched stainless steel interior chamber, HEPA filters and SafeCell™ UV (ultra-violet lamp, integrated in MCO-20AIC but optional for MCO-5AC). MCO-5AC and MCO-15AC utilise Thermal Conductivity (TC) sensors to monitor CO₂ levels whereas MCO-20AIC has unique ceramic-based IR Infrared sensors that deliver accurate CO₂ control with faster recovery after a door opening. Units with user-friendly design, accessories can be easily removed, reversible inner and outer doors, LED digital control and Ø 30 mm access port.
More Product Information