Climate chambers, ICH series
Supplier: MEMMERT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ICH110 ICH110L ICH260 ICH260L ICH260LT01 ICH260LT81 ICH750 ICH750L
390-0663EA 9480 GBP
390-0663 390-0665 390-0666 390-0668 MEMMICH260LT01 MEMMICH260LT81 390-0669 390-0671
Climate chambers, ICH series
Testing Chambers Photostability Chambers
The ICH range of climatic chambers offers rapid and precise temperature control from 10 up to 60 °C with humidification from 10 to 80% RH. These compressor cooled units have stainless steel inner chamber and housing, ensuring long working life and easy cleaning. The ICH models have humidity control, ICH L models feature humidity control and light and ICH C models have humidity and an electronic CO₂ control with automatic zero setting, NDIR measurement system and auto-diagnostic system.
  • Intuitive, user friendly TwinDISPLAY with 2 TFT displays.
More Product Information