Incubators with natural convection, General Protocol Heratherm®
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
51028131 51028132 51028130
390-0468EA 2380 GBP
390-0468 390-0469 390-0467
Incubators with natural convection, General Protocol Heratherm®
Incubators Standard Incubators
Heratherm® General Protocol incubators feature a smooth stainless steel inner chamber with an internal glass door, allowing samples to be viewed without impacting on temperature stability with natural convection providing gentle airflow and minimal drying. These units are recommended for a wide range of routine laboratory applications in pharmaceutical, medical, food and research laboratories and for microbiology and yeast growth. Units have an easy calibration routine and provide enhanced sample safety with an automatic over-temperature alarm to protect samples and incubator.