Cooled Incubators, Series 3, Model 200
Supplier: LMS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
100W 200W 300W 400W 100WA 200WA 300WA 400WA 100WNP 200WNP 300WNP 400WNP
390-0081EA 4580 GBP
390-0081 390-0085 390-0089 390-0096 390-0209 390-0535 390-0211 390-0214 390-0082 390-0086 390-0090 390-0097
Cooled Incubators, Series 3, Model 200
Incubators Cooled Incubators
LMS Series 3 cooled incubators are temperature controlled cabinets with fan assisted air circulation via a pre-mixing chamber. The units have a white enamelled sheet steel exterior, stainless steel lined interior with polyurethane foam insulation, and feature a door lock and magnetic door gasket. The corrosion resistant inner chamber is easy to clean, disinfection procedures are detailed in the user manual.
  • Digital control with digital readout, doorlock
  • Fan assisted air circulation via premixing chamber
  • Full P.I.D. heating/cooling control to ensure stable
More Product Information