Molybdenum standard solution, 10000 mg/L Mo in ammonium hydroxide solution (max. 1%) (from (NH₄)₂MoO₄), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

38726.AE 38726.AP
38726.AEEA 120 GBP
38726.AE 38726.AP
Molybdenum standard solution, 10000 mg/L Mo in ammonium hydroxide solution (max. 1%) (from (NH₄)₂MoO₄), standard for ICP, Specpure®
Standards Cationic Standards Molybdenum Standards

Molecular Formula: (NH₄)₂MoO₄ in 1% NH₄OH
Physical Form: Liquid
Odour: Ammonia-like
MDL No.: MFCD00003465


Certifications: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR