Sodium alginate, viscosity_medium

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Alginic acid sodium salt, Algin

385770025 385770050 385772500 385775000
385770025.EA 276 GBP
385770025. 385770050. 385772500. 385775000.
Sodium alginate, viscosity_medium
Sodium alginate
Storage Temperature: Ambient
MDL Number: MFCD00081310
CAS Number: 9005-38-3
Merck Index: 13,00238

Order Now

Specification Test Results

Appearance almost white to yellow-brown powder
Infrared spectrum Conforms
Loss on drying ≤15.0 % (100 mg, 105°C, 4 hrs)
Heavy metals (as Pb) ≤20 ppm
Sulfated ash 30.0 % to 36.0 % (on dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR