Infrared optical windows
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
38395.KT 38397.KT 38399.KT 38415.KT 38416.KT 38425.KT 38429.KT 38430.KT 38451.KT 38455.KT 38457.KT 38458.KT 38461.KT 38467.KT 38469.KT 38470.KT 38473.KT 38476.KT 38478.KT 38481.KT 45944.KT
38395.KTEA 35.8 GBP
38395.KT 38397.KT 38399.KT 38415.KT 38416.KT 38425.KT 38429.KT 38430.KT 38451.KT 38455.KT 38457.KT 38458.KT 38461.KT 38467.KT 38469.KT 38470.KT 38473.KT 38476.KT 38478.KT 38481.KT 45944.KT
Infrared optical windows
Optical Windows
These optical crystal windows are ideal for infrared spectroscopy.
  • Disc or rectangular
  • Polished or unpolished
A polishing kit is available for those who wish to polish their windows.