Sodium iodide ≥99.5% ACS, Fluka™

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

383112-100G 383112-2.5KG 383112-500G 383112-12KG
383112-100GEA 98.1 GBP
383112-100G 383112-2.5KG 383112-500G 383112-12KG
Sodium iodide ≥99.5% ACS, Fluka™
Sodium iodide
Formula: NaI
Boiling Pt: 1.304 °C (1.013 hPa)
Melting Pt: 662 °C
Density: 3.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003532
CAS Number: 7681-82-5
EINECS: 231-679-3
UN: 3077
ADR: 9,III
Merck Index: 13,08705

Order Now

Specification Test Results

Assay Min. 99.5 %
pH (5 %, 25 °C) 6.0 - 9.0
Water insoluble matter Max. 0.01 %
Barium (Ba) Max. 0.002 %
Calcium (Ca) Max. 0.002 %
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 0.01 %
Magnesium (Mg) Max. 0.001 %
Heavy metals (as Pb) Max. 5 ppm
Chloride and bromide (as Cl) Max. 0.01 %
Iodate (IO₃) Max. 3 ppm
Phosphate (PO₄) Max. 0.001 %
Sulfate (SO₄) Max. 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR