Palladium 20% on activated carbon powder wetted with ca. 50% water, eggshell, unreduced

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

38307.06 38307.14
38307.06EA 301 GBP
38307.06 38307.14
Palladium 20% on activated carbon powder wetted with ca. 50% water, eggshell, unreduced
Palladium

CAS No.: 7440-05-3 Physical Form: 900-1100m2/g Odour: Odourless MDL No.: MFCD03457879


Applications: Debenzylation

Formula: Pd
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR