Potassium iodate concentrate for dilution to 1L, 1/60 M KIO3 (0.1N) Volumetric, Fluka™

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

38120-1EA
38120-1EAEA 85.1 GBP
38120-1EA
Potassium iodate concentrate for dilution to 1L, 1/60 M KIO3 (0.1N) Volumetric, Fluka™
Potassium iodate

Concentrate for 1L standard solution

Formula: KlO₃
MW: 214 g/mol
Boiling Pt: 100 °C
Melting Pt: 560
Storage Temperature: Ambient
MDL Number: MFCD00011406
CAS Number: 7758-05-6
EINECS: 231-831-9
Merck Index: 13,07726

Order Now

Specification Test Results

Factor 0.998 - 1.002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR