Glassy carbon, powder 80-200 µm Type 1

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

38002.09 38002.18 38002.30
38002.09EA 91.2 GBP
38002.09 38002.18 38002.30
Glassy carbon, powder 80-200 µm Type 1
Glassy carbon
Formula: C
Melting Pt: 3652 °C
Density: 1.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
UN: 1325
ADR: 4.1,III
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR