Glassy carbon, rods, Ø 5 mm (0.2 in) Type 2

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

37998.DJ 37998.DM 37998.DR
37998.DJEA 206 GBP
37998.DJ 37998.DM 37998.DR
Glassy carbon, rods, Ø 5 mm (0.2 in) Type 2
Glassy carbon

Physical Form: ≈0.28g/cm MDL No.: MFCD00133992

Formula: C
Melting Pt: 3652 °C
Density: 1.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR