Lithium-tert-butanolate 2.2 M in THF, AcroSeal®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

379311000 379318000
379311000.EA 61 GBP
379311000. 379318000.
Lithium-tert-butanolate 2.2 M in THF, AcroSeal®
Lithium tert-butanolate
Formula: C₄H₉LiO
MW: 80.06 g/mol
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00050479
CAS Number: 1907-33-1
EINECS: 217-611-5
UN: 2924
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear yellow to brown Solution
Strength 19 to 21 %
Impurity ≤0.2 % LiOH + Li2CO3 (as LiOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR