Karl Fischer reagents, buffer for bases, HYDRANAL™
Supplier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
37859-1L 37859-6X1L
37859-1LEA 73.2 GBP
37859-1L 37859-6X1L
Karl Fischer reagents, buffer for bases, HYDRANAL™
Karl Fischer Titrants and Reagents
Liquid buffer medium for Karl Fischer titration in alkaline samples (based on salicylic acid).