Sodium nitrate 99,0, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

3770-01EA 150 GBP
3770-01 3770-07 3770-09
Sodium nitrate 99,0, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Sodium nitrate
Formula: NaNO₃
MW: 84.99 g/mol
Boiling Pt: 380
Melting Pt: 308
Density: 2.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011119
CAS Number: 7631-99-4
EINECS: 231-554-3
UN: 1498
ADR: 5.1 III
REACH: 01-2119488221-41
Merck Index: 13,08720

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements ≥99.0%
For Laboratory,Research,or Manufacturing Use 5.5 - 8.3
ACS - Assay (NaNO₃) ≥99.0 %
ACS - pH of 5% Solution at 25°C 5.5 - 8.3
Insoluble Matter ≤0.005 %
Chloride (Cl) ≤0.001 %
ACS - Iodate (IO₃) ≤5 ppm
ACS- Nitrite (NO₂) ≤0.001 %
ACS - Phosphate (PO₄) ≤5 ppm
ACS - Sulfate (SO₄) ≤0.003 %
Calcium (Ca) ≤0.005 %
ACS - Magnesium (Mg) ≤0.002 %
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Iron (Fe) ≤3 ppm
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR