Sodium cyanotrihydroborate 1M in THF, AcroSeal®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Sodium cyanoborohydride

372451000 372458000
372451000.EA 34.8 GBP
372451000. 372458000.
Sodium cyanotrihydroborate 1M in THF, AcroSeal®
Sodium cyanotrihydroborate
Formula: CH₃BNNa
MW: 62.84 g/mol
Melting Pt: 241…243 °C
Density: 0.915 g/cm³
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00003516
CAS Number: 25895-60-7
UN: 3286
ADR: 3 II
Merck Index: 13,08680

Order Now

Specification Test Results

Appearance Clear to slightly hazy colorless Solution (May develop some turbidity or precipitate)
Titration Iodimetric 0.95 to 1.10 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR