Phenylmercury chloride (63.5% Hg) ≥96%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

37113.09 37113.18
37113.09EA 29.4 GBP
37113.09 37113.18
Phenylmercury chloride (63.5% Hg) ≥96%
Phenylmercury chloride

Purity: 96% CAS No.: 100-56-1 Molecular Formula: C6H5HgCl Formula Weight: 313.18 Physical Form: Powder Appearance: White Melting Point: 246-250° Solubility: Soluble in benzene, ether, pyridine. Very slightly soluble in water Merck Reference: 14,7301 UN No.: 2026 MDL No.: MFCD00000463

Formula: C₆H₅ClHg
Melting Pt: 246…250 °C
Storage Temperature: Ambient
MDL Number: MFCD00000463
CAS Number: 100-56-1
EINECS: 202-865-1
UN: 2026
ADR: 6.1,III
Merck Index: 14,7301

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR