Niobium-n-butoxide ≥99% (metals basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

36570.03 36570.06 36570.14
36570.03EA 48.8 GBP
36570.03 36570.06 36570.14
Niobium-n-butoxide ≥99% (metals basis)
Niobium-n-butoxide

Purity: 99% CAS No.: 51030-47-8 Molecular Formula: C20H45NbO5 Formula Weight: 458.12 Physical Form: Liquid Density: 1.09 Sensitivity: Moisture Sensitive Boiling Point: 197°/0.15mm MDL No.: MFCD00144916

Formula: C₂₀H₄₅NbO₅
MW: 458.12 g/mol
Boiling Pt: 197 °C (0.15 mmHg)
Density: 1.09
Storage Temperature: Ambient
MDL Number: MFCD00144916
CAS Number: 51030-47-8
EINECS: 256-923-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR