Ethyl acetate 99.8%, AcroSeal® for biochemistry

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Acetic acid ethyl ester

364240010 364240025
364240010.EA 36.6 GBP
364240010. 364240025.
Ethyl acetate 99.8%, AcroSeal® for biochemistry
Ethyl acetate
Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 75…78 °C (760 mmHg)
Melting Pt: –83.5 °C
Density: 0.902 g/cm³
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
UN: 1173
ADR: 3 II
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.10 % (Coulometric)
GC ≥99.75 %
UV at 255 nm A: ≤1.0
Color scale ≤10 APHA
Specific gravity (20°C) 0.900 to 0.904
Residue after evaporation ≤0.0002 %
Acidity ≤0.0008 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR