Magnesium acetate tetrahydrate 98.0-102.0% ACS

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Acetic acid magnesium salt tetrahydrate, Magnesium di(acetate) tetrahydrate

36358.22 36358.36 36358.A4 36358.B3
36358.22EA 25.9 GBP
36358.22 36358.36 36358.A4 36358.B3
Magnesium acetate tetrahydrate 98.0-102.0% ACS
Magnesium acetate tetrahydrate

Purity: 98.0-102.0% CAS No.: 16674-78-5 Molecular Formula: Mg(CH3COO)2.4H2O Formula Weight: 214.45 (142.39anhy) Physical Form: Crystalline Density: 1.454 Melting Point: 80° Beilstein Reference: 3730605 Merck Reference: 14,5654 MDL No.: MFCD00149214

Formula: (H₃CCOO)₂Mg·4H₂O
MW: 214.46 g/mol
Melting Pt: 80
Density: 1.454
Storage Temperature: Ambient
MDL Number: MFCD00149214
CAS Number: 16674-78-5
EINECS: 205-554-9
Merck Index: 14,5654

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR