Aluminium potassium sulphate dodecahydrate 98.0-102.0% ACS

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

36288.36 36288.A3
36288.36EA 44.2 GBP
36288.36 36288.A3
Aluminium potassium sulphate dodecahydrate 98.0-102.0% ACS
Aluminium potassium sulphate dodecahydrate

Purity: 98.0-102.0% CAS No.: 7784-24-9 Molecular Formula: KAl(SO4)2.12H2O Formula Weight: 474.38 (258.20anhy) Physical Form: Granular Density: 1.757 Melting Point: 92° Solubility: Soluble in water, glycerol. Insoluble in alcohol Merck Reference: 14,360 MDL No.: MFCD00149143


In printing, fabrics, dyeing, manufacturing of paper, vegetable glue, baking powder, purifying water, clarifying sugar, as catalyst in ammonia synthesis, hardening agent in microscopy

Formula: AlK(SO₄)₂·12H₂O
Melting Pt: 92 °C
Density: 1.757
Storage Temperature: Ambient
MDL Number: MFCD00149143
CAS Number: 7784-24-9
EINECS: 233-141-3
Merck Index: 14,360

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR