Zirconium ≥99.8% (metals basis excluding Hf), lumps 3-6 mm (0.1-0.2 in) (max. 4% Hf)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

36253.14 36253.22
36253.14EA 65.2 GBP
36253.14 36253.22
Zirconium ≥99.8% (metals basis excluding Hf), lumps 3-6 mm (0.1-0.2 in) (max. 4% Hf)
Zirconium

Purity: 99.8% CAS No.: 7440-67-7 Physical Form: For vacuum deposition Odour: Odourless MDL No.: MFCD00011303

Formula: Zr
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR