Crucibles
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
35886.KT 35890.KT 35928.KT 35932.KT 35953.KT 35994.KT 36012.KT 36016.KT 36037.KT 36056.KT 36072.KT 36085.KT 36090.KT 36099.KT
35886.KTEA 64.5 GBP
35886.KT 35890.KT 35928.KT 35932.KT 35953.KT 35994.KT 36012.KT 36016.KT 36037.KT 36056.KT 36072.KT 36085.KT 36090.KT 36099.KT
Crucibles
Crucibles
These crucibles are manufactured from Inconel® nickel-chromium-iron alloy 601. Available in low form and straight wall types.
  • Excellent resistance to oxidation in the 1000 to 1200 °C temperature range
  • Good corrosion resistance to many acid and aqueous salt solutions
  • Unaffected by rapid changes from hot to cold
  • Retains its mechanical strength at elevated temperatures
Designed for applications that require resistance to heat and corrosion. The high resistance of Inconel® alloy 601 to oxidation, carburisation or sulfidation make it well suited forMore Product Information