Crucibles, nickel
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
35883.KT 35889.KT 35904.KT 35906.KT 35925.KT 35927.KT 35946.KT 35948.KT 35952.KT 35967.KT 35991.KT 36040.KT 36053.KT 36071.KT 36088.KT 36098.KT
35883.KTEA 26.5 GBP
35883.KT 35889.KT 35904.KT 35906.KT 35925.KT 35927.KT 35946.KT 35948.KT 35952.KT 35967.KT 35991.KT 36040.KT 36053.KT 36071.KT 36088.KT 36098.KT
Crucibles, nickel
Crucibles
These crucibles are manufactured from nickel. Available in high form, low form and straight wall types.
  • Resistant to phosphoric acid
  • Highly resistant to the corrosive effect of the strongest alkalies
Nickel crucibles offer high resistance to dilute alkalies. In some instances, nickel crucibles are preferable to zirconium: for instance, sodium peroxide fusions in which zirconium itself is to be determined; also in analysis for columbium (niobium), tantalum or low phosphorus.