Strontium ≥99% (metals basis), granules ≤ 19 mm (0.76 in)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


Packaged under argon.

35789.06 35789.14 35789.22
35789.06EA 48.3 GBP
35789.06 35789.14 35789.22
Strontium ≥99% (metals basis), granules ≤ 19 mm (0.76 in)
Strontium

Purity: 99% CAS No.: 7440-24-6 Physical Form: Packaged under argon with thin oil layer Sensitivity: Air and Moisture Sensitive Appearance: White to pale yellow UN No.: 3208 MDL No.: MFCD00134060

Formula: Sr
Boiling Pt: 1151 °C (1013 hPa)
Melting Pt: 570…660 °C
Density: 2.38 g/cm³ (770 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134060
CAS Number: 7440-24-6
EINECS: 231-133-4
UN: 3208
ADR: 4.3,II
Merck Index: 12,08994

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR