Tungsten standard solution, 1000 mg/L W in 5% nitric acid with hydrofluoric acid (max. 1%) (from W), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

35770.AD 35770.AE 35770.AP
35770.ADEA 38.5 GBP
35770.AD 35770.AE 35770.AP
Tungsten standard solution, 1000 mg/L W in 5% nitric acid with hydrofluoric acid (max. 1%) (from W), standard for ICP, Specpure®
Standards Cationic Standards Tungsten Standards

Molecular Formula: W in 5% HNO₃/tr. HF
Physical Form: Liquid
Odour: Acidic
UN No.: 3264
MDL No.: MFCD00011461


Certifications: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR