Yttrium(III) bromide, ultra dry ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


ampouled under argon

35746.04 35746.09
35746.04EA 94 GBP
35746.04 35746.09
Yttrium(III) bromide, ultra dry ≥99.9% (REO, rare earth oxide basis)
Yttrium (III) bromide

Purity: 99.9% CAS No.: 13469-98-2 Molecular Formula: YBr3 Formula Weight: 328.63 Physical Form: Powder, Ampouled under argon Sensitivity: Hygroscopic MDL No.: MFCD00049710

Formula: Br₃Y
Melting Pt: 904 °C
Storage Temperature: Ambient
MDL Number: MFCD00049710
CAS Number: 13469-98-2
EINECS: 236-728-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR