Lactic acid standardised solution 1.0 N in aqueous solution

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

35628.AP
35628.APEA 67.4 GBP
35628.AP
Lactic acid standardised solution 1.0 N in aqueous solution
Lactic acid

CAS No.: 50-21-5 Molecular Formula: C3H6O3 Formula Weight: 90.08 Physical Form: Liquid Appearance: Colourless

Formula: H₃CCH(OH)COOH
Storage Temperature: Ambient
MDL Number: MFCD00004520
CAS Number: 50-21-5
EINECS: 200-018-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR