Sodium chloride standardised solution 0.1 N in aqueous solution

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

35616.AP 35616.K2
35616.APEA 25 GBP
35616.AP 35616.K2
Sodium chloride standardised solution 0.1 N in aqueous solution
Sodium chloride

CAS No.: 7647-14-5 Molecular Formula: NaCl Formula Weight: 58.44 Physical Form: Liquid MDL No.: MFCD00003477

Formula: NaCl
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR