Sodium thiosulphate 1 M Na2S2O3 (1N) volumetric solution, Fluka™

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

35244-1L 35244-6X1L
35244-1LEA 61.3 GBP
35244-1L 35244-6X1L
Sodium thiosulphate 1 M Na2S2O3 (1N) volumetric solution, Fluka™
Sodium thiosulphate
Formula: Na₂S₂O₃
Boiling Pt: 100 °C (1.013 hPa)
Density: 1.120 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5

Order Now

Specification Test Results

Factor 0.999 - 1.001


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR