Copper(II) sulphate 0.1 M CuSO4 Volumetric, Fluka™

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Cupric sulphate

35185-1L 35185-6X1L
35185-1LEA 183 GBP
35185-1L 35185-6X1L
Copper(II) sulphate 0.1 M CuSO4 Volumetric, Fluka™
Copper (II) sulphate
Formula: CuSO₄
MW: 159.61 g/mol
Boiling Pt: >100 °C (1.013 hPa)
Melting Pt: 200
Density: 1.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010981
CAS Number: 7758-98-7
EINECS: 231-847-6
Merck Index: 12,02722

Order Now

Specification Test Results

Factor 0.999 - 1.001


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR