Methyl ethyl ketone ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: EK 500 Additive, Ethyl methyl ketone, 2-Butanone, Butanone, MEK

34861-1L 34861-2.5L 34861-4X2.5L 34861-6X1L 34861-100ML
34861-1LEA 178 GBP
34861-1L 34861-2.5L 34861-4X2.5L 34861-6X1L 34861-100ML
Methyl ethyl ketone ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Methyl ethyl ketone

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency


Beilstein Number: 741880

Formula: C₂H₅COCH₃
MW: 72.11 g/mol
Boiling Pt: 79…81 °C
Melting Pt: –87 °C
Density: 0.805 g/cm³ (20 °C)
Flash Pt: 11.5 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00011648
CAS Number: 78-93-3
EINECS: 201-159-0
UN: 1193
ADR: 3 II
REACH: 01-2119457290-43
Merck Index: 13,06097
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b6656888f6a9a4c0c5ed3053bb2a3dcd8b3af66&vid=17cb98abf677b90f8bac752664d5c0cfd6765591&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.7 %
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.1 %
Free acid (as CH₃COOH) Max. 0.001 %
Transmittance at 335 nm Min. 50 %
Transmittance at 340 nm Min. 80 %
Transmittance from 350 nm Min. 98 %
Explosion Limit 10.1 %
Autoignition Temperature 960 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR