1,4-Dioxane 99,5, CHROMASOLV™ Plus HPLC, Riedel-de Haën™

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide

34857-1L 34857-2.5L 34857-4X2.5L
34857-1LEA 183 GBP
34857-1L 34857-2.5L 34857-4X2.5L
1,4-Dioxane 99,5, CHROMASOLV™ Plus HPLC, Riedel-de Haën™
1,4-Dioxane

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities


Beilstein Number: 102551

Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 100…102 °C (1.013 hPa)
Melting Pt: 12 °C
Density: 1.03 g/cm³ (20 °C)
Flash Pt: 22.5 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00006571
CAS Number: 123-91-1
EINECS: 204-661-8
UN: 1165
ADR: 3 II
Merck Index: 13,03330
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df424642d4a7a96d2eebfb99cf9a2e6d128185d0&vid=37c3b8119f755f99624576b4e975000cbbb8349d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (color) complying
Appearance (form) complying
Vapor Density 3 (vs air)
Purity (GC) Min. 99.50 %
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.05 %
Free acid (as CH₃COOH) Max. 0.002 %
Peroxides (as H₂O₂) Max. 0.01 %
Absorbance at 400 nm Max. 0.01
Absorbance at 300 nm Max. 0.01
Absorbance at 295 nm Max. 0.01
Absorbance at 270 nm Max. 0.10
Absorbance at 250 nm Max. 0.20
Absorbance at 235 nm Max. 0.50
Absorbance at 220 nm Max. 0.70
Transmittance from 220 nm Min. 20 %
Transmittance from 270 nm Min. 80 %
Transmittance from 300 nm Min. 98 %
Vapor Pressure 27 mmHg ( 20 °C)
Autoignition Temperature 356 °F
Explosion Limit 22 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR