Cyclohexane ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

34855-7L 34855-1L 34855-2.5L 34855-4X2.5L 34855-6X1L
34855-7LEA 1210 GBP
34855-7L 34855-1L 34855-2.5L 34855-4X2.5L 34855-6X1L
Cyclohexane ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Cyclohexane

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Formula: C₆H₁₂
Boiling Pt: 81 °C (1.013 hPa)
Melting Pt: 6.4 °C
Density: 0.78 g/cm³ (20 °C)
Flash Pt: 8.3 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) Min. 99.70 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.01 %
Free acid (as HCl) Max. 0.001 %
Absorbance at 400 nm Max. 0.01
Absorbance at 300 nm Max. 0.01
Absorbance at 255 nm Max. 0.01
Absorbance at 250 nm Max. 0.03
Absorbance at 240 nm Max. 0.08
Absorbance at 235 nm Max. 0.10
Absorbance at 230 nm Max. 0.20
Absorbance at 220 nm Max. 0.50
Absorbance at 210 nm Max. 1.00
Fluorescence (chinin) at 254 nm Max. 1.0 ppb
Fluorescence (chinin) at 365 nm Max. 1.0 ppb
APHA Max. 10
Vapor Pressure 77 mmHg ( 20 °C)
Refractive Index n20/D 1.426 (lit.)
Explosion Limit 9 %
Autoignition Temperature 500 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR