Reagents for water determination by the Coulometric Titration Karl Fischer method, Hydranal™
Supplier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
34821-50ML 34836-500ML 34807-500ML 34810-500ML 34820-500ML 34843-500ML 34739-500ML 34829-1L 34840-50ML 34726-500ML 34836-1L 34726-6X500ML 34739-6X500ML 34807-6X500ML 34810-6X500ML 34820-6X500ML 34829-6X1L 34836-6X1L 34836-6X500ML 34843-6X500ML 34821-6X50ML 34840-6X50ML
34821-50MLEA 183.73 GBP
34821-50ML 34836-500ML 34807-500ML 34810-500ML 34820-500ML 34843-500ML 34739-500ML 34829-1L 34840-50ML 34726-500ML 34836-1L 34726-6X500ML 34739-6X500ML 34807-6X500ML 34810-6X500ML 34820-6X500ML 34829-6X1L 34836-6X1L 34836-6X500ML 34843-6X500ML 34821-6X50ML 34840-6X50ML
Reagents for water determination by the Coulometric Titration Karl Fischer method, Hydranal™
Karl Fischer Titrants and Reagents
Coulometric Karl Fischer titrations normally require two reagent solutions: an anolyte (the solution in the anodic compartment) and a catholyte (the solution in the cathodic compartment).
  • Easy to use
  • High accuracy for trace amounts of water
  • Stable conditions of the titration vessel
  • Broad product range available
  • Long shelf life (up to five years)