Petroleum spirit, 40…60 °C, CHROMASOLV™ for pesticide residue analysis, low boiling point hydrogen treated naphtha, Riedel-de Haën™

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

34491-45L 34491-7L 34491-1L 34491-2.5L 34491-4X2.5L
34491-45LEA 3620 GBP
34491-45L 34491-7L 34491-1L 34491-2.5L 34491-4X2.5L
Petroleum spirit, 40…60 °C, CHROMASOLV™ for pesticide residue analysis, low boiling point hydrogen treated naphtha, Riedel-de Haën™
Petroleum spirit

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent

Boiling Pt: 40…60 °C (1.013 hPa)
Density: 0.65 g/cm³ (20 °C)
Flash Pt: –30 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 309-852-0
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.01 %
Suitability for residue analysis Max. 5 ng/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR