Forskolin, Millipore®

Supplier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: (3R,4aR,5S,6S,6aS,10S,10aR,10bS)-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-3-vinyldodecahydro-1H-benzo[f]chromen-5-yl acetate

344270-10 344270-25 344270-50
344270-10EA 207 GBP
344270-10 344270-25 344270-50
Forskolin, Millipore®
Forskolin
Formula: C₂₂H₃₄O₇
MDL Number: MFCD00082317
CAS Number: 66575-29-9
EINECS: 266-410-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR