Anti-Fodrin (Fod009) Mouse Monoclonal Antibody

Supplier: Merck Millipore (Oncogene)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

344050-50
344050-50EA 447 GBP
344050-50
Anti-Fodrin (Fod009) Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Fodrin (Fod009)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR