Bromophenol blue 0,04% ethanol, TECHNICAL

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein

34307.237EA 79.8 GBP
34307.237 34307.237DP
Bromophenol blue 0,04% ethanol, TECHNICAL
Bromophenol blue

  • Transition range: pH 3.0 - 4.6

Formula: C₁₉H₁₀Br₄O₅S
MW: 669.97 g/mol
Melting Pt: 273
Storage Temperature: Ambient
MDL Number: MFCD00005875
CAS Number: 115-39-9
EINECS: 204-086-2
UN: 1170
ADR: 3 II
Merck Index: 13,01429

Order Now

Specification Test Results

Identification Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR