Organic reference standard, Diisobutyl phthalate-3,4,5,6-d4, analytical standard, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

34204-25MG
34204-25MGEA 240 GBP
34204-25MG
Organic reference standard, Diisobutyl phthalate-3,4,5,6-d4, analytical standard, Supelco®
Standards Organic Standards
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=441a0fd44abca3864018d93245d0e91f471e4707&vid=441a0fd44abca3864018d93245d0e91f471e4707&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Description Diisobutyl phthalate-3,4,5,6-d4
Environmentally Preferable
CAS No. 358730-88-8
Formula C₁₆D₄H₁₈O₄

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR